siRNA Details
Gene Symbol: P4HB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000918.3 | NP_000909.2 | 2, 3 | 547 | ||
GenBank mRNA | AK075503.1 | - | 952 | |||
AK095938.1 | - | 2, 3 | 425 | |||
AK296206.1 | - | 2, 3 | 414 | |||
AK297081.1 | - | 2, 3 | 444 | |||
AK297967.1 | - | 1, 2 | 367 | |||
AK315631.1 | - | 2, 3 | 378 | |||
AU131354.1 | - | 2, 3 | 546 | |||
BC010859.1 | - | 2, 3 | 394 | 3879411 | ||
BC029617.1 | - | 2, 3 | 394 | 3848651 | ||
BC071892.1 | - | 2, 3 | 353 | 5443660 | ||
J02783.1 | - | 2, 3 | 413 | |||
X05130.1 | - | 2, 3 | 379 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
