siRNA Details
Gene Symbol: CSNK1G1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022048.3 | NP_071331.2 | 7, 8 | 1253 | ||
GenBank mRNA | AB042562.1 | - | 6, 7 | 901 | ||
AB042563.1 | - | 6, 7 | 885 | |||
AF223357.2 | - | 6, 7 | 971 | |||
AK026957.1 | - | 3, 4 | 360 | |||
AK225960.1 | - | 7, 8 | 1183 | |||
AL832551.1 | - | 7, 8 | 1031 | |||
BC017236.2 | - | 6, 7 | 702 | |||
BC130613.1 | - | 7, 8 | 1026 | 40146644 | ||
BC136504.1 | - | 7, 8 | 1026 | 9020493 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
