siRNA Details
Gene Symbol: BACH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001186.3 | NP_001177.1 | 5 | 3556 | ||
NM_206866.2 | NP_996749.1 | 5 | 3580 | |||
GenBank mRNA | AB002803.1 | - | 5 | 3455 | ||
BC063307.1 | - | 5 | 3429 | 30340811 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
