siRNA Details
Gene Symbol: GP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007240.2 | NP_001007241.2 | 3 | 496 | ||
NM_001502.3 | NP_001493.2 | 3 | 496 | |||
GenBank mRNA | AB035541.1 | - | 2 | 285 | ||
AK312277.1 | - | 3 | 383 | |||
BX646730.1 | - | 3 | 390 | |||
BX647831.1 | - | 3 | 384 | |||
BX648023.1 | - | 3 | 390 | |||
CR749600.1 | - | 3 | 455 | |||
DA041213.1 | - | 3 | 496 | |||
U36221.1 | - | 3 | 365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
