siRNA Details
Gene Symbol: ATP5F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001688.4 | NP_001679.2 | 3 | 583 | ||
GenBank mRNA | AK223018.1 | - | 3 | 208 | ||
AK291077.1 | - | 3 | 240 | |||
AK308266.1 | - | 4 | 619 | |||
AK313541.1 | - | 3 | 210 | |||
AY871272.1 | - | 3 | 217 | |||
BC005366.1 | - | 3 | 228 | 3839745 | ||
BC005960.1 | - | 3 | 220 | 4049491 | ||
BC016350.1 | - | 3 | 259 | 4074170 | ||
X60221.1 | - | 3 | 194 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
