siRNA Details
Gene Symbol: CRYZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130042.1 | NP_001123514.1 | 10 | 1443 | ||
NM_001130043.1 | NP_001123515.1 | 8 | 1302 | |||
NM_001134759.1 | NP_001128231.1 | 7 | 1116 | |||
NM_001889.3 | NP_001880.2 | 9 | 1404 | |||
XM_005270491.4 | XP_005270548.1 | 9 | 917 | |||
XM_011540747.1 | XP_011539049.1 | 9 | 1023 | |||
XM_017000367.1 | XP_016855856.1 | 9 | 976 | |||
XM_017000368.1 | XP_016855857.1 | 8 | 927 | |||
GenBank mRNA | AB209714.1 | - | 10 | 1075 | ||
AK123469.1 | - | 2 | 1484 | |||
AK128794.1 | - | 9 | 1009 | |||
AK223150.1 | - | 9 | 1036 | |||
AK223201.1 | - | 9 | 1036 | |||
AK314813.1 | - | 8 | 942 | |||
BC039578.1 | - | 9 | 1026 | 6067125 | ||
BC070058.1 | - | 10 | 1085 | 5267616 | ||
BX647883.1 | - | 7 | 727 | |||
BX649061.1 | - | 2 | 2037 | |||
L13278.1 | - | 8 | 948 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
