siRNA Details
Gene Symbol: DCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001129889.2 | NP_001123361.1 | 10 | 2158 | ||
NM_001322182.1 | NP_001309111.1 | 7 | 1281 | |||
NM_001322183.1 | NP_001309112.1 | 8 | 1335 | |||
NM_001322184.1 | NP_001309113.1 | 9 | 1635 | |||
NM_001322185.1 | NP_001309114.1 | 8 | 1581 | |||
NM_001322186.1 | NP_001309115.1 | 10 | 1850 | |||
NM_001922.4 | NP_001913.2 | 8 | 2059 | |||
XM_011521049.1 | XP_011519351.1 | 10 | 2683 | |||
XM_017020401.1 | XP_016875890.1 | 10 | 2191 | |||
GenBank mRNA | AJ132932.1 | - | 2 | 453 | ||
AK293115.1 | - | 8 | 1842 | |||
BC028311.1 | - | 8 | 1906 | 4705935 | ||
D17547.1 | - | 8 | 2046 | |||
DQ902581.1 | - | 10 | 2023 | |||
L18967.1 | - | 8 | 2016 | |||
S69231.1 | - | 8 | 1981 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
