siRNA Details
Gene Symbol: CSN1S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001890.1 | NP_001881.1 | 6, 7 | 209 | ||
XM_006714090.2 | XP_006714153.1 | 6, 7 | 228 | |||
GenBank mRNA | BC128227.1 | - | 6, 7 | 198 | 40114618 | |
BC128228.1 | - | 6, 7 | 198 | 40114622 | ||
U23157.1 | - | 6, 7 | 188 | |||
X78416.1 | - | 6, 7 | 209 | |||
X98084.1 | - | 5, 6 | 137 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
