siRNA Details
Gene Symbol: LEPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002303.5 | NP_002294.2 | 20 | 3998 | ||
GenBank mRNA | U66497.1 | - | 19 | 3939 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
