siRNA Details
Gene Symbol: LSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145436.1 | NP_001138908.1 | 22 | 2559 | ||
NM_001145437.1 | NP_001138909.1 | 21 | 2724 | |||
NM_002340.5 | NP_002331.3 | 22 | 2592 | |||
XM_011529564.2 | XP_011527866.1 | 22 | 2618 | |||
XM_011529566.2 | XP_011527868.1 | 22 | 2539 | |||
XM_011529567.2 | XP_011527869.1 | 21 | 2522 | |||
GenBank mRNA | AK092334.1 | - | 3 | 1942 | ||
AK098352.1 | - | 2 | 78 | |||
AK128839.1 | - | 2303 | ||||
AK226141.1 | - | 21 | 2637 | |||
BC035638.1 | - | 22 | 2555 | 5547173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
