siRNA Details
Gene Symbol: LY9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261456.1 | NP_001248385.1 | 5 | 1204 | ||
NM_002348.3 | NP_002339.2 | 5 | 1204 | |||
XM_011509548.1 | XP_011507850.1 | 6 | 1264 | |||
XM_011509549.1 | XP_011507851.1 | 6 | 1264 | |||
XM_011509550.1 | XP_011507852.1 | 6 | 1264 | |||
XM_011509552.1 | XP_011507854.1 | 6 | 1264 | |||
XM_017001297.1 | XP_016856786.1 | 6 | 1264 | |||
XM_017001298.1 | XP_016856787.1 | 6 | 1264 | |||
XM_017001299.1 | XP_016856788.1 | 6 | 1264 | |||
XM_017001300.1 | XP_016856789.1 | 5 | 1233 | |||
XM_017001301.1 | XP_016856790.1 | 5 | 1104 | |||
XM_017001304.1 | XP_016856793.1 | 5 | 1159 | |||
GenBank mRNA | AF244129.1 | - | 5 | 1117 | ||
AK292048.1 | - | 5 | 1104 | |||
BC062589.1 | - | 2 | 257 | |||
L42621.1 | - | 5 | 997 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
