siRNA Details
Gene Symbol: CTSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257971.1 | NP_001244900.1 | 3 | 460 | ||
NM_001257972.1 | NP_001244901.1 | 3 | 401 | |||
NM_001912.4 | NP_001903.1 | 3 | 550 | |||
NM_145918.2 | NP_666023.1 | 3 | 405 | |||
XM_005251716.3 | XP_005251773.1 | 4 | 1280 | |||
XM_011518263.1 | XP_011516565.1 | 3 | 1090 | |||
XM_017014293.1 | XP_016869782.1 | 3 | 1027 | |||
GenBank mRNA | AK055599.1 | - | 3 | 1088 | ||
AK075100.1 | - | 3 | 356 | |||
AL832167.1 | - | 3 | 348 | |||
BC012612.1 | - | 3 | 398 | 4295635 | ||
BC142983.1 | - | 3 | 345 | 8860422 | ||
BI824449.1 | - | 3 | 399 | |||
BI829385.1 | - | 3 | 504 | |||
BX537395.1 | - | 3 | 341 | |||
BX647101.1 | - | 3 | 345 | |||
BX647102.1 | - | 3 | 344 | |||
BX647413.1 | - | 3 | 345 | |||
BX647434.1 | - | 3 | 403 | |||
BX647435.1 | - | 3 | 400 | |||
BX648848.1 | - | 3 | 355 | |||
BX648849.1 | - | 3 | 345 | |||
BX649140.1 | - | 3 | 401 | |||
CR457053.1 | - | 2 | 200 | |||
DN993009.1 | - | 2 | 420 | |||
X12451.1 | - | 3 | 488 |
Gene Symbol: CTSL3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027917.1 | 1 | 35 | |||
GenBank mRNA | AJ851862.1 | - | 1 | 35 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
