siRNA Details
Gene Symbol: OGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165036.1 | NP_001158508.1 | 11 | 1652 | ||
NM_002541.3 | NP_002532.2 | 11 | 1664 | |||
XM_005249759.4 | XP_005249816.1 | 12 | 1641 | |||
XM_005249761.3 | XP_005249818.1 | 12 | 1647 | |||
XM_011515408.2 | XP_011513710.1 | 13 | 1728 | |||
GenBank mRNA | AK293871.1 | - | 11 | 1420 | ||
AK294951.1 | - | 8 | 1038 | |||
AK296400.1 | - | 8 | 1165 | |||
AK302805.1 | - | 9 | 1276 | |||
AK304439.1 | - | 11 | 1583 | |||
AK304690.1 | - | 9 | 1247 | |||
AK314179.1 | - | 11 | 1586 | |||
BC004964.1 | - | 10 | 1547 | 3528538 | ||
BC014617.2 | - | 10 | 1547 | 3010095 | ||
BC128549.1 | - | 10 | 1211 | |||
BC128550.1 | - | 10 | 1223 | |||
D10523.1 | - | 11 | 1562 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
