siRNA Details
Gene Symbol: OGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165036.1 | NP_001158508.1 | 14 | 1988 | ||
NM_002541.3 | NP_002532.2 | 14 | 2000 | |||
XM_005249759.4 | XP_005249816.1 | 15 | 1977 | |||
XM_005249761.3 | XP_005249818.1 | 15 | 1983 | |||
XM_011515408.2 | XP_011513710.1 | 16 | 2064 | |||
GenBank mRNA | AK293871.1 | - | 14 | 1756 | ||
AK294951.1 | - | 11 | 1374 | |||
AK296400.1 | - | 11 | 1501 | |||
AK302805.1 | - | 12 | 1612 | |||
AK304439.1 | - | 14 | 1919 | |||
AK304690.1 | - | 12 | 1583 | |||
AK314179.1 | - | 14 | 1922 | |||
BC004964.1 | - | 13 | 1883 | 3528538 | ||
BC014617.2 | - | 13 | 1883 | 3010095 | ||
BC128549.1 | - | 13 | 1547 | |||
BC128550.1 | - | 13 | 1559 | |||
D10523.1 | - | 14 | 1898 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
