siRNA Details
Gene Symbol: OGDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165036.1 | NP_001158508.1 | 23 | 4194 | ||
NM_002541.3 | NP_002532.2 | 23 | 4206 | |||
XM_005249759.4 | XP_005249816.1 | 24 | 4183 | |||
XM_005249761.3 | XP_005249818.1 | 24 | 4189 | |||
XM_011515408.2 | XP_011513710.1 | 25 | 4270 | |||
GenBank mRNA | BC004964.1 | - | 22 | 4089 | 3528538 | |
BC014617.2 | - | 22 | 4089 | 3010095 | ||
D10523.1 | - | 23 | 4102 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
