siRNA Details
Gene Symbol: ZNF143 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282656.1 | NP_001269585.1 | 16 | 2415 | ||
NM_001282657.1 | NP_001269586.1 | 15 | 2325 | |||
NM_003442.5 | NP_003433.3 | 16 | 2418 | |||
XM_005253121.1 | XP_005253178.1 | 16 | 2366 | |||
XM_005253122.1 | XP_005253179.1 | 16 | 2391 | |||
XM_005253127.1 | XP_005253184.1 | 17 | 2444 | |||
XM_011520349.2 | XP_011518651.1 | 16 | 3137 | |||
XM_017018252.1 | XP_016873741.1 | 15 | 2302 | |||
XM_017018253.1 | XP_016873742.1 | 15 | 2299 | |||
XM_017018254.1 | XP_016873743.1 | 16 | 2403 | |||
XM_017018255.1 | XP_016873744.1 | 16 | 2493 | |||
XM_017018256.1 | XP_016873745.1 | 15 | 2305 | |||
GenBank mRNA | AF071771.1 | - | 13 | 2038 | ||
BC020219.1 | - | 16 | 2417 | 4902668 | ||
CN295530.1 | - | 1 | 464 |
