siRNA Details
Gene Symbol: ZNF202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301779.1 | NP_001288708.1 | 8 | 4211 | ||
NM_001301780.1 | NP_001288709.1 | 7 | 4067 | |||
NM_001301819.1 | NP_001288748.1 | 6 | 3928 | |||
NM_003455.3 | NP_003446.2 | 9 | 4263 | |||
GenBank mRNA | AF027218.1 | - | 6 | 3866 | ||
AF027219.1 | - | 6 | 3870 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
