siRNA Details
Gene Symbol: UNC5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003728.3 | NP_003719.3 | 10, 11 | 2079 | ||
XM_005263321.3 | XP_005263378.1 | 11, 12 | 2136 | |||
XM_017008789.1 | XP_016864278.1 | 11, 12 | 8496 | |||
XM_017008790.1 | XP_016864279.1 | 10, 11 | 1724 | |||
GenBank mRNA | AF055634.1 | - | 10, 11 | 1880 | ||
AK290500.1 | - | 10, 11 | 1890 | |||
AK308421.1 | - | 11, 12 | 1899 |
