siRNA Details
Gene Symbol: MBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003927.4 | NP_003918.1 | 4 | 1094 | ||
GenBank mRNA | AF039701.1 | - | 3 | 232 | ||
AF072242.1 | - | 4 | 1094 | |||
BC032638.1 | - | 4 | 935 | 5496721 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
