siRNA Details
Gene Symbol: MAP3K19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005007.1 | XP_016860496.1 | 9 | 4644 | ||
GenBank mRNA | AK026727.1 | - | 1 | 1424 | ||
AK225579.1 | - | 1 | 1424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
