siRNA Details
Gene Symbol: ZNF235 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004234.4 | NP_004225.3 | 5 | 852 | ||
XM_005259412.4 | XP_005259469.1 | 5 | 4908 | |||
GenBank mRNA | AK226126.1 | - | 1 | 1881 | ||
AK300317.1 | - | 5 | 852 | |||
AK315242.1 | - | 5 | 827 | |||
BC117215.1 | - | 5 | 825 | 40125766 | ||
BC143482.1 | - | 5 | 825 |
