siRNA Details
Gene Symbol: ZNHIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281433.1 | NP_001268362.1 | 5 | 650 | ||
NM_001281434.1 | NP_001268363.1 | 3 | 516 | |||
NM_004773.3 | NP_004764.1 | 5 | 684 | |||
NR_104009.1 | 5 | 679 | ||||
NR_104010.1 | 4 | 652 | ||||
GenBank mRNA | AB209193.1 | - | 3 | 4148 | ||
AF400652.1 | - | 5 | 676 | |||
BC017931.1 | - | 5 | 649 | 4669658 | ||
BC105608.1 | - | 4 | 624 | |||
BE260293.1 | - | 3 | 414 | |||
L40410.1 | - | 5 | 602 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
