siRNA Details
Gene Symbol: PPID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005038.2 | NP_005029.1 | 4 | 599 | ||
GenBank mRNA | AK313929.1 | - | 4 | 600 | ||
BC030707.2 | - | 4 | 595 | 4819748 | ||
CR542081.1 | - | 4 | 487 | |||
L11667.1 | - | 4 | 586 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
