siRNA Details
Gene Symbol: CYB5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001914.3 | NP_001905.1 | 4, 5 | 461 | ||
GenBank mRNA | M60174.1 | - | 4, 5 | 320 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
