siRNA Details
Gene Symbol: TP53BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031685.2 | NP_001026855.2 | 12 | 2029 | ||
NM_005426.2 | NP_005417.1 | 13 | 2161 | |||
XM_011544268.1 | XP_011542570.1 | 11 | 1890 | |||
XM_011544269.2 | XP_011542571.1 | 9 | 1548 | |||
XM_017002220.1 | XP_016857709.1 | 13 | 2170 | |||
GenBank mRNA | AJ318888.2 | - | 12 | 1974 | ||
AK294432.1 | - | 12 | 1881 | |||
AK294964.1 | - | 5 | 1049 | |||
AK295378.1 | - | 6 | 753 | |||
AK316016.1 | - | 5 | 951 | |||
AK316103.1 | - | 5 | 929 | |||
BC040247.1 | - | 10 | 1505 | 4454266 | ||
BC046150.1 | - | 12 | 1972 | |||
BC058918.1 | - | 12 | 1987 | 4537206 | ||
U58334.1 | - | 13 | 2106 |
