siRNA Details
Gene Symbol: ABCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320032.1 | NP_001306961.1 | 11, 12 | 2070 | ||
NM_005688.3 | NP_005679.2 | 11, 12 | 1955 | |||
XM_005247058.4 | XP_005247115.1 | 11, 12 | 3523 | |||
XM_005247059.4 | XP_005247116.1 | 11, 12 | 1945 | |||
XM_011512314.1 | XP_011510616.1 | 11, 12 | 1846 | |||
XM_011512315.1 | XP_011510617.1 | 11, 12 | 2005 | |||
XM_017005492.1 | XP_016860981.1 | 8, 9 | 3194 | |||
GenBank mRNA | AB005659.1 | - | 5, 6 | 1032 | ||
AB019002.1 | - | 10, 11 | 1821 | |||
AB209103.1 | - | 8, 9 | 1465 | |||
AB209454.1 | - | 11, 12 | 1882 | |||
AF104942.1 | - | 11, 12 | 1894 | |||
AF146074.1 | - | 11, 12 | 1965 | |||
AY196484.1 | - | 10, 11 | 1781 | |||
BC140771.1 | - | 10, 11 | 1831 | 9021639 | ||
BE085682.1 | - | 215 | ||||
U83661.2 | - | 11, 12 | 1888 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
