siRNA Details
Gene Symbol: ABCC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320032.1 | NP_001306961.1 | 30 | 4865 | ||
NM_005688.3 | NP_005679.2 | 30 | 4750 | |||
XM_005247058.4 | XP_005247115.1 | 30 | 6318 | |||
XM_005247059.4 | XP_005247116.1 | 30 | 4740 | |||
XM_011512314.1 | XP_011510616.1 | 30 | 4641 | |||
XM_017005492.1 | XP_016860981.1 | 27 | 5989 | |||
GenBank mRNA | AB005659.1 | - | 24 | 3827 | ||
AB019002.1 | - | 29 | 4616 | |||
AB209103.1 | - | 27 | 4259 | |||
AB209454.1 | - | 30 | 4677 | |||
AF104942.1 | - | 30 | 4689 | |||
AF146074.1 | - | 30 | 4760 | |||
BC140771.1 | - | 29 | 4626 | 9021639 | ||
BX647245.1 | - | 12 | 1875 | |||
U83661.2 | - | 30 | 4683 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
