siRNA Details
Gene Symbol: ZNF263 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005741.4 | NP_005732.2 | 6 | 2614 | ||
XM_006720831.1 | XP_006720894.1 | 6 | 2682 | |||
XM_006720832.1 | XP_006720895.1 | 5 | 2604 | |||
XM_006720833.1 | XP_006720896.1 | 4 | 2466 | |||
XM_006720834.3 | XP_006720897.1 | 5 | 2737 | |||
GenBank mRNA | AK295350.1 | - | 2 | 1480 | ||
BC008805.2 | - | 6 | 2612 | 3945548 | ||
D88827.1 | - | 6 | 2574 |
