siRNA Details
Gene Symbol: CAPRIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005898.4 | NP_005889.3 | 19 | 3223 | ||
XM_017017749.1 | XP_016873238.1 | 18 | 3194 | |||
GenBank mRNA | BC001731.1 | - | 19 | 3223 | 3355481 | |
BC015700.1 | - | 5 | 1399 | |||
BC026302.1 | - | 1 | 623 | |||
Z48042.1 | - | 18 | 3076 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
