siRNA Details
Gene Symbol: TRIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282985.1 | NP_001269914.1 | 3 | 2282 | ||
NM_025195.3 | NP_079471.1 | 3 | 3108 | |||
GenBank mRNA | AF205437.1 | - | 3 | 2799 | ||
AF250310.1 | - | 3 | 2802 | |||
AK001319.1 | - | 1 | 886 | |||
AK022787.1 | - | 1 | 886 | |||
AL832388.1 | - | 3 | 2740 | |||
BC063292.1 | - | 3 | 3108 | 30343492 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
