siRNA Details
Gene Symbol: MTHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005957.4 | NP_005948.3 | 6 | 1080 | ||
XM_005263458.3 | XP_005263515.1 | 6 | 1917 | |||
XM_005263460.4 | XP_005263517.1 | 6 | 1535 | |||
XM_005263462.4 | XP_005263519.1 | 6 | 968 | |||
XM_005263463.3 | XP_005263520.1 | 6 | 1097 | |||
XM_011541495.2 | XP_011539797.1 | 6 | 1914 | |||
XM_011541496.2 | XP_011539798.1 | 6 | 1917 | |||
XM_017001328.1 | XP_016856817.1 | 6 | 1917 | |||
GenBank mRNA | AB209113.1 | - | 5 | 1118 | ||
AJ237672.1 | - | 5 | 980 | |||
AK312907.1 | - | 6 | 941 | |||
BC053509.1 | - | 6 | 902 | 6374885 | ||
CN355286.1 | - | 2 | 213 | |||
JQ429447.1 | - | 2 | 356 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
