siRNA Details
Gene Symbol: SCML2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006089.2 | NP_006080.1 | 12, 13 | 1735 | ||
NR_033717.1 | 12, 13 | 1735 | ||||
XM_006724458.1 | XP_006724521.1 | 12, 13 | 1699 | |||
XM_017029219.1 | XP_016884708.1 | 12, 13 | 1631 | |||
XM_017029220.1 | XP_016884709.1 | 12, 13 | 1627 | |||
XM_017029221.1 | XP_016884710.1 | 12, 13 | 1769 | |||
XM_017029222.1 | XP_016884711.1 | 6, 7 | 1013 | |||
GenBank mRNA | AK022824.1 | - | 1, 2 | 135 | ||
AK299190.1 | - | 6, 7 | 1013 | |||
AK303064.1 | - | 11, 12 | 1601 | |||
AK308140.1 | - | 6, 7 | 1009 | |||
AK315116.1 | - | 12, 13 | 1699 | |||
BC051913.1 | - | 12, 13 | 1677 | |||
BC064617.1 | - | 12, 13 | 1692 | 5575894 | ||
Y18004.1 | - | 12, 13 | 1669 |
