siRNA Details
Gene Symbol: PHYH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037537.1 | NP_001032626.1 | 7, 8 | 1164 | ||
NM_001323080.1 | NP_001310009.1 | 8, 9 | 1223 | |||
NM_001323082.1 | NP_001310011.1 | 8, 9 | 1065 | |||
NM_001323083.1 | NP_001310012.1 | 6, 7 | 795 | |||
NM_001323084.1 | NP_001310013.1 | 7, 8 | 1170 | |||
NM_006214.3 | NP_006205.1 | 8, 9 | 1059 | |||
GenBank mRNA | AF023462.1 | - | 8, 9 | 998 | ||
AF112977.1 | - | 8, 9 | 1030 | |||
BC029512.1 | - | 8, 9 | 1019 | 5277185 | ||
CR542055.1 | - | 8, 9 | 971 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
