siRNA Details
Gene Symbol: SLC30A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF006621.2 | - | 3 | 408 | ||
AK225470.1 | - | 3 | 371 | |||
AK299917.1 | - | 3 | 410 | |||
AK312838.1 | - | 3 | 434 | |||
AY319413.1 | - | 4 | 313 | |||
BC000240.1 | - | 3 | 395 | |||
BC007732.2 | - | 3 | 395 | 4109702 | ||
BC016949.1 | - | 3 | 413 | 3850521 | ||
BG323207.1 | - | 434 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
