siRNA Details
Gene Symbol: PDIA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006810.3 | NP_006801.1 | 8 | 724 | ||
NR_028444.1 | 8 | 724 | ||||
GenBank mRNA | BC001625.1 | - | 8 | 671 | 3537659 | |
BM549365.1 | - | 8 | 724 | |||
D49490.1 | - | 8 | 624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
