siRNA Details
Gene Symbol: PYCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282279.1 | NP_001269208.1 | 7 | 1595 | ||
NM_001282280.1 | NP_001269209.1 | 8 | 1688 | |||
NM_001282281.1 | NP_001269210.1 | 8 | 1681 | |||
NM_006907.3 | NP_008838.2 | 7 | 2017 | |||
NM_153824.2 | NP_722546.1 | 8 | 1674 | |||
XM_005256381.2 | XP_005256438.1 | 8 | 1646 | |||
XM_011523583.2 | XP_011521885.1 | 8 | 1885 | |||
XM_011523584.2 | XP_011521886.1 | 8 | 1640 | |||
XM_011523585.2 | XP_011521887.1 | 7 | 1511 | |||
XM_017024909.1 | XP_016880398.1 | 7 | 1524 | |||
GenBank mRNA | AF218000.1 | - | 8 | 1510 | ||
AK225879.1 | - | 7 | 1563 | |||
BC001504.2 | - | 7 | 1614 | 3505512 | ||
BC022244.1 | - | 7 | 1535 | 4420238 | ||
BC071842.1 | - | 7 | 1636 | 5088067 | ||
M77836.1 | - | 7 | 1585 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
