siRNA Details
Gene Symbol: AMACR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167595.1 | NP_001161067.1 | 3 | 638 | ||
NM_014324.5 | NP_055139.4 | 3 | 638 | |||
GenBank mRNA | AF158378.1 | - | 3 | 607 | ||
AJ130733.1 | - | 3 | 630 | |||
AK000912.1 | - | 3 | 623 | |||
AK022765.1 | - | 3 | 608 | |||
AK222831.1 | - | 3 | 610 | |||
AK225749.1 | - | 3 | 647 | |||
AL545355.3 | - | 3 | 551 | |||
AY330489.1 | - | 3 | 551 | |||
AY935981.1 | - | 3 | 551 | |||
BQ941482.1 | - | 1 | 147 | |||
EF560721.1 | - | 3 | 561 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
