siRNA Details
Gene Symbol: TAF5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014409.3 | NP_055224.1 | 5 | 2142 | ||
XM_005273099.4 | XP_005273156.1 | 5 | 2095 | |||
XM_005273100.2 | XP_005273157.1 | 6 | 2108 | |||
XM_011544162.1 | XP_011542464.1 | 4 | 1879 | |||
XM_011544163.1 | XP_011542465.1 | 8 | 2724 | |||
XM_011544164.2 | XP_011542466.1 | 4 | 1822 | |||
XM_017001045.1 | XP_016856534.1 | 7 | 2879 | |||
GenBank mRNA | AJ009770.1 | - | 1 | 989 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
