siRNA Details
Gene Symbol: MICAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282663.1 | NP_001269592.1 | 8, 9 | 1241 | ||
NM_001282664.1 | NP_001269593.1 | 7, 8 | 1094 | |||
NM_001282665.1 | NP_001269594.1 | 7, 8 | 1094 | |||
NM_001282666.1 | NP_001269595.1 | 6, 7 | 1019 | |||
NM_001282667.1 | NP_001269596.1 | 6, 7 | 1019 | |||
NM_014632.3 | NP_055447.1 | 8, 9 | 1245 | |||
XM_011520490.2 | XP_011518792.1 | 6, 7 | 948 | |||
XM_011520491.2 | XP_011518793.1 | 6, 7 | 948 | |||
XM_017018589.1 | XP_016874078.1 | 7, 8 | 1061 | |||
XM_017018590.1 | XP_016874079.1 | 7, 8 | 1106 | |||
XM_017018591.1 | XP_016874080.1 | 7, 8 | 1061 | |||
GenBank mRNA | AB018293.1 | - | 8, 9 | 1206 | ||
AB110785.1 | - | 6, 7 | 942 | |||
AB110786.1 | - | 6, 7 | 942 | |||
AK294845.1 | - | 7, 8 | 1061 | |||
AK302893.1 | - | 8, 9 | 1323 | |||
BC044577.1 | - | 8, 9 | 1230 | 5275364 | ||
BX538021.1 | - | 4, 5 | 769 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
