siRNA Details
Gene Symbol: IPO13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014652.3 | NP_055467.3 | 20 | 3902 | ||
GenBank mRNA | AB018267.1 | - | 20 | 3769 | ||
AK023441.1 | - | 14 | 1625 | |||
BX116797.1 | - | 3 | 530 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
