siRNA Details
Gene Symbol: SV2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167580.1 | NP_001161052.1 | 12 | 3222 | ||
NM_001323031.1 | NP_001309960.1 | 13 | 3755 | |||
NM_001323032.1 | NP_001309961.1 | 13 | 4064 | |||
NM_001323033.1 | NP_001309962.1 | 11 | 3731 | |||
NM_001323034.1 | NP_001309963.1 | 12 | 3863 | |||
NM_001323036.1 | NP_001309965.1 | 12 | 3041 | |||
NM_001323037.1 | NP_001309966.1 | 14 | 3971 | |||
NM_001323038.1 | NP_001309967.1 | 13 | 3883 | |||
NM_001323039.1 | NP_001309968.1 | 13 | 4156 | |||
NM_001323040.1 | NP_001309969.1 | 13 | 3402 | |||
NM_014848.5 | NP_055663.1 | 14 | 4152 | |||
XM_005254998.3 | XP_005255055.1 | 13 | 3817 | |||
XM_017022761.1 | XP_016878250.1 | 13 | 3768 | |||
XM_017022762.1 | XP_016878251.1 | 13 | 3903 | |||
GenBank mRNA | AB018278.1 | - | 12 | 3425 | ||
AL110287.1 | - | 1 | 864 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
