siRNA Details
Gene Symbol: C1orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098616.2 | NP_001092086.1 | 3, 4 | 674 | ||
NM_001297718.1 | NP_001284647.1 | 3, 4 | 597 | |||
NM_001297721.1 | NP_001284650.1 | 2, 3 | 572 | |||
NM_001297723.1 | NP_001284652.1 | 3, 4 | 674 | |||
NM_015449.3 | NP_056264.1 | 2, 3 | 572 | |||
GenBank mRNA | AF077036.1 | - | 2, 3 | 392 | ||
AF109185.1 | - | 3, 4 | 365 | |||
AF529365.1 | - | 3, 4 | 284 | |||
AJ243664.1 | - | 2, 3 | 340 | |||
AJ243665.1 | - | 2, 3 | 177 | |||
AK290583.1 | - | 3, 4 | 446 | |||
AL117463.1 | - | 1, 2 | 138 | |||
BC000152.2 | - | 3, 4 | 483 | 2901006 | ||
BC001377.2 | - | 2, 3 | 340 | 3051048 | ||
BC006514.1 | - | 2, 3 | 409 | 2822327 | ||
BC008306.1 | - | 3, 4 | 410 | 3502049 | ||
BC014391.1 | - | 2, 3 | 308 | 3544590 | ||
BG478941.1 | - | 3, 4 | 404 | |||
CD247026.1 | - | 2, 3 | 417 | |||
CR457131.1 | - | 2, 3 | 182 | |||
DQ908899.1 | - | 2, 3 | 182 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
