siRNA Details
Gene Symbol: POLR2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018102.2 | NP_001018112.1 | 4 | 1349 | ||
NM_015532.4 | NP_056347.1 | 4 | 1820 | |||
GenBank mRNA | AK074955.1 | - | 4 | 1622 | ||
AK128618.1 | - | 4 | 1145 | |||
AK223571.1 | - | 4 | 1619 | |||
AL050091.1 | - | 3 | 988 |
Gene Symbol: GCOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018090.6 | NP_001018100.1 | 14 | 2175 | ||
NM_001018091.6 | NP_001018101.1 | 13 | 1970 | |||
NM_001285900.3 | NP_001272829.1 | 15 | 2820 | |||
NR_104367.2 | 16 | 2954 | ||||
NR_104368.2 | 14 | 2148 | ||||
NR_104369.2 | 13 | 2074 | ||||
NR_104370.2 | 12 | 1990 | ||||
NR_104371.3 | 17 | 3610 | ||||
GenBank mRNA | BC050322.1 | - | 1 | 20 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
