siRNA Details
Gene Symbol: POLR2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018102.2 | NP_001018112.1 | 4 | 2178 | ||
NM_015532.4 | NP_056347.1 | 4 | 2649 | |||
GenBank mRNA | AK074955.1 | - | 4 | 2452 | ||
AK128618.1 | - | 4 | 1973 | |||
AL050091.1 | - | 3 | 1817 |
Gene Symbol: GCOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018090.6 | NP_001018100.1 | 14 | 3004 | ||
NM_001018091.6 | NP_001018101.1 | 13 | 2799 | |||
NM_001285900.3 | NP_001272829.1 | 15 | 3649 | |||
NR_104367.2 | 16 | 3783 | ||||
NR_104368.2 | 14 | 2977 | ||||
NR_104369.2 | 13 | 2903 | ||||
NR_104370.2 | 12 | 2819 | ||||
NR_104371.3 | 17 | 4439 | ||||
GenBank mRNA | AY620236.1 | - | 1 | 191 | ||
BC050322.1 | - | 1 | 849 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
