siRNA Details
Gene Symbol: PPIL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016059.4 | NP_057143.1 | 4 | 743 | ||
GenBank mRNA | AF151882.1 | - | 4 | 718 | ||
AK026636.1 | - | 4 | 545 | |||
AY359032.1 | - | 4 | 720 | |||
BC003048.1 | - | 4 | 695 | 3543388 | ||
BU195579.1 | - | 4 | 744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
