siRNA Details
Gene Symbol: KLF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016270.3 | NP_057354.1 | 3 | 1584 | ||
GenBank mRNA | AF134053.1 | - | 3 | 1486 | ||
AF205849.1 | - | 3 | 1570 | |||
AK225820.1 | - | 3 | 1583 | |||
BC071983.1 | - | 3 | 1573 | 5480246 |
