siRNA Details
Gene Symbol: NKRF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173487.1 | NP_001166958.1 | 4 | 3344 | ||
NM_001173488.1 | NP_001166959.1 | 2 | 3730 | |||
NM_017544.3 | NP_060014.2 | 3 | 3246 | |||
XM_011531365.2 | XP_011529667.1 | 4 | 3876 | |||
GenBank mRNA | AJ011812.2 | - | 2 | 3709 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
