siRNA Details
Gene Symbol: FOXRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017547.3 | NP_060017.1 | 8, 9 | 1146 | ||
NR_037647.1 | 7, 8 | 925 | ||||
NR_037648.1 | 8, 9 | 1165 | ||||
XM_006718879.3 | XP_006718942.1 | 7, 8 | 859 | |||
XM_006718881.3 | XP_006718944.1 | 6, 7 | 827 | |||
XM_011542895.2 | XP_011541197.1 | 8, 9 | 1243 | |||
XM_011542896.2 | XP_011541198.1 | 8, 9 | 1052 | |||
XM_017018000.1 | XP_016873489.1 | 8, 9 | 1121 | |||
XM_017018001.1 | XP_016873490.1 | 8, 9 | 1265 | |||
XM_017018002.1 | XP_016873491.1 | 7, 8 | 774 | |||
XM_017018003.1 | XP_016873492.1 | 7, 8 | 1026 | |||
XM_017018004.1 | XP_016873493.1 | 7, 8 | 1148 | |||
XM_017018005.1 | XP_016873494.1 | 6, 7 | 2381 | |||
XM_017018006.1 | XP_016873495.1 | 7, 8 | 1026 | |||
GenBank mRNA | AF103801.1 | - | 8, 9 | 1044 | ||
AF447877.1 | - | 6, 7 | 1298 | |||
AK023987.1 | - | 8, 9 | 1015 | |||
AK126067.1 | - | 2, 3 | 2124 | |||
AK295267.1 | - | 8, 9 | 1101 | |||
AK295428.1 | - | 8, 9 | 1031 | |||
AK298010.1 | - | 6, 7 | 699 | |||
AK298807.1 | - | 8, 9 | 1031 | |||
AK302039.1 | - | 7, 8 | 794 | |||
AL136923.1 | - | 8, 9 | 1026 | |||
BC002910.2 | - | 6, 7 | 623 | |||
BC013902.2 | - | 8, 9 | 1002 | 3956972 |
