siRNA Details
Gene Symbol: MIOX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017584.5 | NP_060054.4 | 8, 9 | 758 | ||
XM_005261925.3 | XP_005261982.1 | 7, 8 | 625 | |||
GenBank mRNA | AF197129.1 | - | 7, 8 | 644 | ||
AY064416.1 | - | 8, 9 | 671 | |||
AY738258.1 | - | 7, 8 | 644 | |||
BC064395.1 | - | 8, 9 | 666 | |||
BC073848.1 | - | 8, 9 | 678 | 5186354 | ||
CR456478.1 | - | 8, 9 | 703 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
