siRNA Details
Gene Symbol: PHF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018288.3 | NP_060758.2 | 10 | 1206 | ||
NM_133325.2 | NP_579866.2 | 10 | 1200 | |||
XM_017010998.1 | XP_016866487.1 | 9 | 1089 | |||
GenBank mRNA | AK001837.1 | - | 8 | 870 | ||
AK222612.1 | - | 8 | 865 | |||
BC020954.2 | - | 8 | 897 | 3845815 | ||
BC110323.1 | - | 10 | 1177 | 5547708 | ||
CR457273.1 | - | 8 | 863 | |||
KC839988.1 | - | 10 | 1186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
